Learn More
Fisherbrand™ Non-sterile Transfer Pipettes, Transparent LDPE
Prevent cross contamination. Fisherbrand™ Non-sterile Transfer Pipets, Transparent LDPE are made of shatterproof, flexible plastic and are designed to replace glass Pasteur pipets. Graduated and non-graduated pipets available in a range of sizes and styles, including jumbo, thin stem and extra long.
Brand:  Fisherbrand™ 13499108
 View more versions of this product
Product Code. 13499108
  • 20.23 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Built-in bulb eliminates the need for plugging, packaging and autoclaving
  • Flexible stem bends to sample from narrow or small tubes
  • Low affinity surface reduces loss of cells and valuable proteins due to binding
  • Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
  • Non-sterile, transparent and fully recyclable
Specifications
Specifications
LDPE
Yes
500 Pack
33 μL drop
150 mm
30
0.25 mL
Standard
1 mL
Transfer Pipet
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Non-sterile Transfer Pipettes, Transparent LDPE > Capacity: 1mL; Drops/mL: 30; Graduated