missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Transparent Polypropylene Beaker

Use these polypropylene beakers for a variety of general laboratory applications. VITLAB™ Transparent Polypropylene Beakers feature a stable handle easy-grip handle and an easily readable, embossed blue scale.

Brand:  Vitlab™ 441081

 View more versions of this product

Product Code. 10523822

  • 49.50 € / Pack of 12
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Highly transparent polypropylene beaker
  • To preserve markings, do not clean at temperatures exceeding 60°C
  • Conditionally autoclavable at 121°C (2 bar) according to DIN EN 285
  • For autoclaving, we recommend model 440941 with raised graduations
Specifications

Specifications

91 mm
500 mL
Polypropylene
133 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vitlab™ Transparent Polypropylene Beaker > Capacity: 500mL; Height: 133mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.