missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Transparent Polypropylene Powder Funnels

Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.

Brand:  Kartell™ 0016600

5.90 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10524471

  • 12.95 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.

  • 60° angled sides
  • External ribbing avoids pressure lock
  • Can be used with liquids
  • Autoclavable
Specifications

Specifications

180mm
43mm
60°
Can be used with liquids
1290 mL
Polypropylene
Autoclavable
48mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers