Learn More
IKA™ Trayster Basics
GREENER_CHOICE
Trayster Basic Overhead shaker with vertical rotation for smooth but effective mixing with a fixed speed.
Brand:  IKA™ 0004005000
967.43 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 13417669
  • 1069.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for biological samples, particularly blood samples.
  • Suitable for separating and mixing processes of powdered and liquid samples in Eppendorf tubes and Greiner tubes up to 50 mL.
  • The easy removable attachments allow simple and quick cleaning of the unit of spilled samples.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0 to 20 rpm
100/240 V
1 A
50/60 Hz
460 mm
80%
100/240 V, 24 VDC, 50/60Hz, 1A
4°C to 50°C
IP 21 DIN EN 60529
140 mm
Trayster Basic
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ Trayster Basics > Trayster Basic