missing translation for 'onlineSavingsMsg'
Learn More
Tri(1-aziridinyl)phosphine sulfide, 98%, Thermo Scientific™ Product Code.: 15458348

Tri(1-aziridinyl)phosphine sulfide, 98%, Thermo Scientific™

Product Code. 15458348
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15458348

Brand: Thermo Scientific Alfa Aesar H61778.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in benzene, acetone and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-24-4
Molecular Formula C6H12N3PS
Molecular Weight (g/mol) 189.22
MDL Number MFCD00145452
InChI Key FOCVUCIESVLUNU-UHFFFAOYSA-N
Synonym thiotepa, thio-tepa, triethylenethiophosphoramide, thiophosphamide, tiofosfamid, tiofosyl, thiofozil, tespamin, thioplex, tiofozil
PubChem CID 5453
ChEBI CHEBI:9570
IUPAC Name tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane
SMILES S=P(N1CC1)(N1CC1)N1CC1

Specifications

Quantity 1 g
UN Number UN2811
Solubility Information Soluble in benzene,acetone and methanol.
Formula Weight 189.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Tri(1-aziridinyl)phosphine sulfide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tri(1-aziridinyl)phosphine sulfide, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.