missing translation for 'onlineSavingsMsg'
Learn More

β-Tri-Calcium phosphate, ≥98% β to phase basis, Honeywell Fluka™

Product Code. 15665700
Click to view available options
Quantity:
10 g
100 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a.,≥98% β-phase basis (sintered Powder)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-87-4
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.174
MDL Number MFCD00015984
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
PubChem CID 24456
ChEBI CHEBI:9679
IUPAC Name tricalcium;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Melting Point 1670°C
Color White
Physical Form Solid
Quantity 100 g
UN Number NONH for all modes of transport
Formula Weight 310.18g/mol
Percent Purity ≥98% β to phase basis (sintered Powder)
Packaging Poly Bottle
Density 3.27g/cm3 at 20.5°C
Chemical Name or Material β-tri-Calcium phosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.