missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-butylgermanium hydride, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Tri-n-butylgermanium hydride is used in the reduction of benzylic chlorides. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Especificaciones
Color | Colorless to Yellow |
Density | 0.92 g/mL |
Boiling Point | 123°C (20 mmHg) |
Flash Point | 113°C (235°F) |
Refractive Index | 1.4508 |
Quantity | 1 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | Not miscible. |
Formula Weight | 244.95 |
Percent Purity | ≥97% |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'