Learn More
Tri-n-butyltin chloride, 96%, Thermo Scientific Chemicals
27.64 € - 246.23 €
Chemical Identifiers
CAS | 1461-22-9 |
---|---|
Molecular Formula | C12H27ClSn |
Molecular Weight (g/mol) | 325.508 |
MDL Number | MFCD00000521 |
InChI Key | GCTFWCDSFPMHHS-UHFFFAOYSA-M |
Synonym | tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride |
PubChem CID | 15096 |
ChEBI | CHEBI:79734 |
IUPAC Name | tributyl(chloro)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429632
|
Thermo Scientific Alfa Aesar
A10746.18 |
50 g |
27.64 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439632
|
Thermo Scientific Alfa Aesar
A10746.30 |
250 g |
86.43 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419632
|
Thermo Scientific Alfa Aesar
A10746.0B |
1000 g |
246.23 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.
Solubility
Miscible with alcohol, heptane, benzene and toluene. Immiscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
1461-22-9 | |
325.508 | |
GCTFWCDSFPMHHS-UHFFFAOYSA-M | |
15096 | |
tributyl(chloro)stannane |
C12H27ClSn | |
MFCD00000521 | |
tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride | |
CHEBI:79734 | |
CCCC[Sn](CCCC)(CCCC)Cl |
Specifications
1461-22-9 | |
1.2 | |
120°C (248°F) | |
1.491 | |
UN2788 | |
tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride | |
GCTFWCDSFPMHHS-UHFFFAOYSA-M | |
tributyl(chloro)stannane | |
15096 | |
325.51 | |
Tri-n-butyltin chloride |
-19°C | |
171°C to 173°C (25 mmHg) | |
C12H27ClSn | |
MFCD00000521 | |
3535715 | |
Miscible with alcohol,heptane,benzene and toluene. Immiscible with water. | |
CCCC[Sn](CCCC)(CCCC)Cl | |
325.508 | |
CHEBI:79734 | |
96% |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H360FD-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 215-958-7
missing translation for 'rtecsNumber' : WH6820000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only