missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals

Product Code. 11443900
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-52-3
Molecular Formula C13H30OSn
Molecular Weight (g/mol) 321.092
MDL Number MFCD00009419
InChI Key KJGLZJQPMKQFIK-UHFFFAOYSA-N
Synonym tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl
PubChem CID 16683411
IUPAC Name tributyl(methoxy)stannane
SMILES CCCC[Sn](CCCC)(CCCC)OC

Specifications

Density 1.12
Boiling Point 124°C to 126°C (4 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.474
Quantity 100 g
UN Number UN2788
Beilstein 3600960
Sensitivity Moisture sensitive
Formula Weight 321.09
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin methoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.