missing translation for 'onlineSavingsMsg'
Learn More
Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals Product Code.: 11458913

Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals

Product Code. 11458913
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11458913

Brand: Thermo Scientific Alfa Aesar A11062.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

Solubility
Soluble in chloroform, toluene and hexane. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-50-2
Molecular Formula C24H51OP
Molecular Weight (g/mol) 386.645
MDL Number MFCD00002083
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
PubChem CID 65577
IUPAC Name 1-dioctylphosphoryloctane
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 52°C to 57°C
Density 0.88
Boiling Point 212°C to 213°C (0.3 mmHg)
Flash Point 252°C (486°F)
Odor Odorless
Quantity 500 g
Beilstein 1796648
Solubility Information Soluble in chloroform,toluene and hexane. Insoluble in water.
Formula Weight 386.65
Percent Purity 98%
Chemical Name or Material Tri-n-octylphosphine oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252893 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog NumberA11062.14,A11062.36,A11062.22
Lot Number10251946 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberA11062.14,A11062.36,A11062.22
Lot Number10242927 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog NumberA11062.14,A11062.36,A11062.22
Lot Number10240264 Certificate TypeCertificate of Analysis Date25/04/2022 Catalog NumberA11062.14,A11062.36,A11062.22
Lot Number10239892 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11062.14,A11062.36,A11062.22
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.