Learn More
Tri-n-propyltin chloride, Thermo Scientific Chemicals
47.60 € - 763.80 €
Chemical Identifiers
CAS | 2279-76-7 |
---|---|
Molecular Formula | C9H21ClSn |
Molecular Weight (g/mol) | 283.427 |
MDL Number | MFCD00013610 |
InChI Key | FVFLIQMADUVDSP-UHFFFAOYSA-M |
Synonym | tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride |
PubChem CID | 16784 |
IUPAC Name | chloro(tripropyl)stannane |
SMILES | CCC[Sn](CCC)(CCC)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394809
|
Thermo Scientific Alfa Aesar
071122.03 |
1 g | N/A | ||||||
11384809
|
Thermo Scientific Alfa Aesar
071122.06 |
5 g | N/A | ||||||
11374809
|
Thermo Scientific Alfa Aesar
071122.14 |
25 g |
763.80 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2279-76-7 | |
283.427 | |
FVFLIQMADUVDSP-UHFFFAOYSA-M | |
16784 | |
CCC[Sn](CCC)(CCC)Cl |
C9H21ClSn | |
MFCD00013610 | |
tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride | |
chloro(tripropyl)stannane |
Specifications
2279-76-7 | |
1.2678 g/mL at 28°C | |
C9H21ClSn | |
MFCD00013610 | |
Moisture sensitive | |
FVFLIQMADUVDSP-UHFFFAOYSA-M | |
chloro(tripropyl)stannane | |
16784 | |
Liquid |
-23.5°C | |
123°C (13 mmHg) | |
1.491 | |
UN2788 | |
tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride | |
CCC[Sn](CCC)(CCC)Cl | |
283.427 | |
283.41 | |
Tri-n-propyltin chloride |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 218-910-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.