Learn More
Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent,
Brand:  Thermo Scientific™ TS-49001
 View more versions of this product
Product Code. 11587841
  • 245.22 € / Each
Explore more special offers
Description
Description
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent, Composed of HMDS, TMCS and high-purity pyridine, it rapidly produces TMS derivatives of polar compounds for GC analysis and biochemical synthesis. This versatile reagent is ideally suited for GC determinations of a range of compounds including sugars, alcohols, phenols, steroids, sterols, organic acids, and some amines.
  • Derivatizes carbohydrates, phenols, steroids, sterols, organic acids, alcohols and some amines
  • Useful for rapid production of TMS derivatives of polar compounds for gas chromatographic determination and biochemical synthesis
  • Versatility, speed and ease of use of Tri-Suil HTP Reagent has made it the most widely used silylation formulation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GC
65 - 80, 10 - 20, 5 - 7%
2°C to 8°C
Pyridine, Hexamethyldisilazane, Chlorotrimethylsilane
Corrosive, Flammable
50 mL
50 mL