Learn More
Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent.
Marke: Thermo Scientific™ TS-49230
307.97 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Beschreibung
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent. Comprising TMSI in pyridine, this reagent silylates alcohols and phenols, organic acids, hydroxylamines, amino acids, carbohydrates, flavonoids, glycols and polyglycols, nucleotides, steroids, hydroxyl acids, barbituates, narcotics, indoles, and vitamins.
Additonal Product InformationDoes not react with amines
May be used in the presence of water as long as there is enough reagent present to react with both the water and the sample
Spezifikation
GC | |
65 - 80, 20 - 25% | |
2°C to 8°C | |
Pyridine, 1-(Trimethylsilyl)-1 H-imidazole |
Harmful | |
10 x 1 mL ampules | |
10 x 1 mL ampules |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only. Not for use in diagnostic procedures.