missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tri-Sodium citrate dihydrate, BAKER, J.T. Baker™
Click to view available options
1 kg
Specifications
Specifications
Melting Point | 150°C |
CAS | 6132-04-3 |
Quantity | 1 kg |
Assay Percent Range | 99.0 to 100.5% |
Molecular Formula | C6H9Na3O9 |
MDL Number | MFCD00150031 |
Synonym | Sodium Citrate, Dihydrate , Citric acid trisodium salt dihydrate , Sodium Citrate Dihydrate , Sodium citrate tribasic dihydrate |
InChI Key | NLJMYIDDQXHKNR-UHFFFAOYSA-K |
SMILES | O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
IUPAC Name | trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction