Learn More
Tocris Bioscience™ Triacsin C
Inhibitor of acyl-CoA synthetase
Brand:  Tocris Bioscience™ 2472/100U
184.02 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18778598
  • 238.00 € / 100µg
Explore more special offers
Description
Description
Triacsin C is an inhibitor of acyl-CoA synthetase. Inhibits lipid synthesis and cell proliferation in Raji lymphoma cells. Induces apoptosis in cancer cell lines and inhibits human lung cancer cell xenograft tumor growth in mice.
Specifications
Specifications
>95%
Triacsin C
Triacsin C
100 μg
207.27