Learn More
BRAND™ Triangular Wedge-Shape PTFE Magnetic Stirring Bars
An excellent option for liquid agitation in the laboratory.
Brand: BRAND™ 137815
65.37 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
- Ideal for dissolving solids and mixing of sediments due to scraping action
- Angular surface creates high turbulence even at low speeds
- Provided in packs of 10
Specifications
Triangular wedge-shaped | |
35 mm | |
10/Pk. |
10 mm | |
PTFE | |
Triangle |