Learn More
BRAND™ Triangular Wedge-Shape PTFE Magnetic Stirring Bars
An excellent option for liquid agitation in the laboratory.
Brand:  BRAND™ 137810
52.08 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12095282
  • 79.50 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for dissolving solids and mixing of sediments due to scraping action
  • Angular surface creates high turbulence even at low speeds
  • Provided in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Triangular wedge-shaped
25 mm
10/Pk.
8 mm
PTFE
Triangle