Learn More
Tribenzyltin chloride, Thermo Scientific™
26.60 € - 362.00 €
Chemical Identifiers
CAS | 3151-41-5 |
---|---|
Molecular Formula | C21H21ClSn |
Molecular Weight (g/mol) | 427.559 |
MDL Number | MFCD00014431 |
InChI Key | PUFMEHZUXSPPAM-UHFFFAOYSA-M |
Synonym | tribenzyltin chloride, tribenzylchlorotin, chlorotribenzyltin, stannane, chlorotribenzyl, chlorid tribenzylcinicity, stannane, tribenzylchloro, chlorotribenzylstannane, tribenzylstannium chloride, tin, tribenzyl-, chloride, stannane, chlorotris phenylmethyl |
PubChem CID | 18470 |
IUPAC Name | tribenzyl(chloro)stannane |
SMILES | C1=CC=C(C=C1)C[Sn](CC2=CC=CC=C2)(CC3=CC=CC=C3)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3151-41-5 | |
427.559 | |
PUFMEHZUXSPPAM-UHFFFAOYSA-M | |
18470 | |
C1=CC=C(C=C1)C[Sn](CC2=CC=CC=C2)(CC3=CC=CC=C3)Cl |
C21H21ClSn | |
MFCD00014431 | |
tribenzyltin chloride, tribenzylchlorotin, chlorotribenzyltin, stannane, chlorotribenzyl, chlorid tribenzylcinicity, stannane, tribenzylchloro, chlorotribenzylstannane, tribenzylstannium chloride, tin, tribenzyl-, chloride, stannane, chlorotris phenylmethyl | |
tribenzyl(chloro)stannane |
Specifications
3151-41-5 | |
C21H21ClSn | |
UN3146 | |
tribenzyltin chloride, tribenzylchlorotin, chlorotribenzyltin, stannane, chlorotribenzyl, chlorid tribenzylcinicity, stannane, tribenzylchloro, chlorotribenzylstannane, tribenzylstannium chloride, tin, tribenzyl-, chloride, stannane, chlorotris phenylmethyl | |
C1=CC=C(C=C1)C[Sn](CC2=CC=CC=C2)(CC3=CC=CC=C3)Cl | |
427.559 | |
427.54 | |
Tribenzyltin chloride |
143°C | |
MFCD00014431 | |
Moisture sensitive | |
PUFMEHZUXSPPAM-UHFFFAOYSA-M | |
tribenzyl(chloro)stannane | |
18470 | |
Powder |
Safety and Handling
GHS H Statement
H301-H319-H335
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c
H301-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures