Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Product Code.: 11975141
Product Code. 11975141
10 L , Metal drum
Click to view available options
Packaging:
Glass Bottle
Metal drum
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: According to the Federal Republic of Germany's Export Control Regulations and the German Chemical Companies Association (VCI), we must obtain a customer declaration for each customer before supplying
Product Code.
11975141
Brand: Thermo Scientific Acros
169930100
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: According to the Federal Republic of Germany's Export Control Regulations and the German Chemical Companies Association (VCI), we must obtain a customer declaration for each customer before supplying
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
126-73-8
Molecular Formula
C12H27O4P
Molecular Weight (g/mol)
266.32
MDL Number
MFCD00009436
InChI Key
STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym
tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de Show More Show Less
PubChem CID
31357
ChEBI
CHEBI:35019
IUPAC Name
tributyl phosphate
SMILES
CCCCOP(=O)(OCCCC)OCCCC
Specifications
Melting Point
-79.0°C
Density
0.9790g/mL
Boiling Point
289.0°C
Flash Point
146°C
Infrared Spectrum
Authentic
Assay Percent Range
99% min. (GC)
Packaging
Metal drum
Linear Formula
[CH3 (CH2 )3 O]3 P(O)
Refractive Index
1.4230 to 1.4250
Quantity
10 L
Beilstein
01,II,397
Merck Index
15,9781
Specific Gravity
0.979
Solubility Information
Solubility in water: 0.6g/100mL (20°C). Other solubilities: soluble in alcohols,ketones,ethers,benzene,,carbon disulfide and most organic solvents
Viscosity
3.5 mPa.s (25°C)
Formula Weight
266.32
Percent Purity
99+%
Physical Form
Liquid
Chemical Name or Material
Tributyl phosphate, 99%
Show More
Show Less
You may be interested in these alternatives
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number
Certificate Type
Certificate of Analysis
Certificate of Compliance
Certificate of Sterility/Irradiation
Certificate of Validation
Certificate of Guarantee
Certificate of Animal Origin
Certificate of Quality
Certificate of Processing
Declaration of Conformity
Search
5 results found
Lot Number
Certificate Type
Date
Catalog Number
Lot Number A0468027
Certificate Type Certificate of Analysis
Date 31/01/2025
Catalog Number 169930025,169930010,169930100
Lot Number A0466616
Certificate Type Certificate of Analysis
Date 07/10/2024
Catalog Number 169930025,169932500,169930010,169930100
Lot Number A0465091
Certificate Type Certificate of Analysis
Date 01/08/2024
Catalog Number 169930025,169932500,169930010,169930100
Lot Number A0460570
Certificate Type Certificate of Analysis
Date 04/03/2024
Catalog Number 169930025,169932500,169930010,169930100
Lot Number A0460569
Certificate Type Certificate of Analysis
Date 04/03/2024
Catalog Number 169930025,169932500,169930010,169930100
Product Identifier
Signal Word
Hazard Category
Acute toxicity Category 4
LONG-TERM AQUATIC HAZARD Chronic 3
Carcinogenicity Category 2
Skin corrosion/irritation Category 2
Hazard Statement
H315-Causes skin irritation.
H351-Suspected of causing cancer.
H412-Harmful to aquatic life with long lasting effects.
H302-Harmful if swallowed.
Precautionary Statement
P201-Obtain special instructions before use.
P280-Wear protective gloves/protective clothing/eye protection/face protection.
P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
P302+P352-IF ON SKIN: Wash with plenty of water/.
P312-Call a POISON CENTER/doctor/if you feel unwell.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tributyl phosphate, 99+%, Thermo Scientific Chemicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.