missing translation for 'onlineSavingsMsg'
Learn More

Tributylamine, ≥98.5%, Honeywell™

Product Code. 15648410
Click to view available options
Quantity:
25 mL
250 mL
2 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

≥98.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-82-9
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009431
InChI Key IMFACGCPASFAPR-UHFFFAOYSA-N
Synonym tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7
PubChem CID 7622
ChEBI CHEBI:38905
IUPAC Name N,N-dibutylbutan-1-amine
SMILES CCCCN(CCCC)CCCC

Specifications

Melting Point -70°C
Color Colorless
Density 0.778g/mL at 25°C
Boiling Point 216°C
Flash Point 63°C (145.4°F)
Odor Unpleasant
Packaging Glass Bottle
Linear Formula [CH3(CH2)3]3N
Quantity 25 mL
UN Number UN2542
Beilstein 1698872
Formula Weight 185.35g/mol
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Tributylamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'