missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylamine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -70.0°C |
Density | 0.7780g/mL |
Boiling Point | 215.0°C |
Flash Point | 75°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | [CH3NCH2)3]3N |
Refractive Index | 1.4273 to 1.4293 |
Quantity | 100 mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0469056 | Certificate of Analysis | 31/01/2025 | 139320100,139321000,139320010,139322500,139320025 |
A0436193 | Certificate of Analysis | 11/11/2024 | 139320100,139321000,139320010,139322500,139320025 |
A0441367 | Certificate of Analysis | 20/10/2023 | 139320100,139321000,139320010,139322500,139320025 |
A0441366 | Certificate of Analysis | 20/10/2023 | 139320100,139321000,139320010,139322500,139320025 |
A0436794 | Certificate of Analysis | 25/08/2023 | 139320100,139321000,139320010,139322500,139320025 |
Safety and Handling

Product Identifier
- Tributylamine
- Danger
- Acute toxicity Category 1
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H315-Causes skin irritation.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tributylamine, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction