missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyrin, 97%, Thermo Scientific Chemicals
Tributyrin, 97%, C15H26O6, CAS Number-60-01-5, butyrin, butyryl triglyceride, glycerol tributanoate, glycerol tributyrate, glyceroltributyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, tributin, tributyrin, tri-n-butyrin, 100mL, Liquid, 96% min. (GC), 02, 273, 305.0 deg.C to 310.0 deg.C
30.50 € - 128.00 €
Chemical Identifiers
CAS | 60-01-5 |
---|---|
Molecular Formula | C15H26O6 |
Molecular Weight (g/mol) | 302.36 |
MDL Number | MFCD00009392 |
InChI Key | UYXTWWCETRIEDR-UHFFFAOYSA-N |
Synonym | tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin |
PubChem CID | 6050 |
ChEBI | CHEBI:35020 |
IUPAC Name | 2,3-di(butanoyloxy)propyl butanoate |
SMILES | CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10387750
|
Thermo Scientific Acros
150881000 |
100 mL | Glass bottle |
30.50 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10163340
|
Thermo Scientific Acros
150882500 |
250 mL | Glass bottle |
44.70 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10554264
|
Thermo Scientific Acros
150880010 |
1 L | Glass bottle |
128.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-01-5 | |
302.36 | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
6050 | |
2,3-di(butanoyloxy)propyl butanoate |
C15H26O6 | |
MFCD00009392 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
CHEBI:35020 | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Specifications
60-01-5 | |
Clear colorless to light yellow | |
305.0°C to 310.0°C | |
Conforms | |
C15H26O6 | |
1.4340 to 1.437 | |
02, 273 | |
1.0335 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
302.36 | |
CHEBI:35020 | |
97% | |
Tributyrin, 98% |
-75.0°C | |
1.0335g/mL | |
173°C | |
96% min. (GC) | |
(CH3CH2CH2CO2CH2)2CH(O2CCH2CH2CH3) | |
MFCD00009392 | |
15, 9785 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
2,3-di(butanoyloxy)propyl butanoate | |
6050 | |
302.36 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 200-451-5
missing translation for 'rtecsNumber' : ET7350000
missing translation for 'tsca' : TSCA
RUO – Research Use Only