missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricine, Electrophoresis Grade, MP Biomedicals
5704-04-1 Purity: >>>99% Heavy metals (as Pb): <<10 ppm pKa at 25°C = 8.1 Biological buffer
136.00 € - 885.00 €
Chemical Identifiers
CAS | 5704-04-1 |
---|---|
Molecular Formula | C6H13NO5 |
Molecular Weight (g/mol) | 179.17 |
InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
PubChem CID | 79784 |
ChEBI | CHEBI:39063 |
IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
SMILES | C(C(=O)O)NC(CO)(CO)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11431632
|
MP Biomedicals
|
100 g |
136.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441632
|
MP Biomedicals
04807416 |
250 g |
273.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11451632
|
MP Biomedicals
|
1 kg |
885.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
5704-04-1 | |
179.17 | |
79784 | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
C6H13NO5 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
CHEBI:39063 | |
C(C(=O)O)NC(CO)(CO)CO |
Specifications
5704-04-1 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
79784 | |
Electrophoresis |
C6H13NO5 | |
C(C(=O)O)NC(CO)(CO)CO | |
179.17 | |
CHEBI:39063 | |
Tricine |