Learn More
Cytiva Tricorn Columns
Tricorn columns are compatible with high flow rates and are designed for high-performance media such as Sepharose, Superdex, and SOURCE.
Brand:  Cytiva 28-4064-13
697.30 EUR valid until 2024-12-31
Use promo code "21963" to get your promotional price.
 View more versions of this product
Product Code. 10640571
  • 734.00 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tricorn empty columns are designed for high performance separations on small and medium scale
  • The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
  • Broad range of column dimensions suitable for many different purification needs.
  • Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
  • Separate packing connector allows simple column packing for a broad range of media.
  • Locking ring prevents accidental compression of media bed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tricorn Columns (Empty)
10mm
20mm
Borosilicate glass
Polypropylene filter
High resolution liquid chromatography of biomolecules performed at medium to high pressures
0.00-2.29mL (one adapter), 0.00-2.07mL (two adapters)
1 Ea.
Glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock and filter holder
725psi (50 bar)
Tricorn Column (Empty)