Learn More
Tricyclohexylphosphine, 98%, Thermo Scientific Chemicals
112.00 € - 351.00 €
Chemical Identifiers
CAS | 2622-14-2 |
---|---|
Molecular Formula | C18H33P |
Molecular Weight (g/mol) | 280.436 |
MDL Number | MFCD00003853 |
InChI Key | WLPUWLXVBWGYMZ-UHFFFAOYSA-N |
Synonym | tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration |
PubChem CID | 75806 |
IUPAC Name | tricyclohexylphosphane |
SMILES | C1CCC(CC1)P(C2CCCCC2)C3CCCCC3 |
Description
Used as an active pharmaceutical ingredient. Ligand used in the Pd-catalyzed coupling of malononitrile with aryl halides. Bulky phosphine ligand used with a Pd(0)-triolefinic macrocycle catalyst for Suzuki coupling of aryl bromides and chlorides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2622-14-2 | |
280.436 | |
WLPUWLXVBWGYMZ-UHFFFAOYSA-N | |
75806 | |
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3 |
C18H33P | |
MFCD00003853 | |
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration | |
tricyclohexylphosphane |
Specifications
2622-14-2 | |
White | |
C18H33P | |
Air Sensitive | |
Insoluble in water. | |
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3 | |
280.436 | |
280.42 | |
Crystalline |
76°C to 78°C | |
Unpleasant | |
MFCD00003853 | |
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration | |
WLPUWLXVBWGYMZ-UHFFFAOYSA-N | |
tricyclohexylphosphane | |
75806 | |
98% | |
Tricyclohexylphosphine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 220-069-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only