missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, ∽99%, MP Biomedicals™

Product Code. 11493501
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 102-71-6
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO

Specifications

Color White
Assay Percent Range ∽ 99%
Molecular Formula C6H15NO3
Quantity 1 kg
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Molecular Weight (g/mol) 149.19
Formula Weight 185.65
Physical Form Powder
Chemical Name or Material Triethanolamine Hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'