missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals

Product Code. 11407627
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Protein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-39-8
Molecular Formula C6H16ClNO3
Molecular Weight (g/mol) 185.648
MDL Number MFCD00012596
InChI Key HHLJUSLZGFYWKW-UHFFFAOYSA-N
Synonym triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy
PubChem CID 101814
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
SMILES C(CO)N(CCO)CCO.Cl

Specifications

Melting Point 177°C to 179°C
Linear Formula (HOCH2CH2)3N·HCl
Quantity 250 g
Beilstein 3909940
Sensitivity Hygroscopic
Merck Index 14,9665
Solubility Information Soluble in water.
Formula Weight 185.65
Percent Purity ≥99%
Chemical Name or Material Triethanolamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'