missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99.7%, extra pure, Thermo Scientific Chemicals

Product Code. 10438643
Click to view available options
Quantity:
5 g
50 g
1 kg
2.5 kg
10 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115.0°C
Density 0.7280g/mL
Boiling Point 90.0°C
Flash Point −11°C
Infrared Spectrum Authentic
Assay Percent Range 99.6% min. (GC)
Packaging Metal Drum
Molecular Formula C6H15N
Linear Formula (C2H5)3N
Refractive Index 1.3990 to 1.4010
Quantity 10 kg
Beilstein 04, 99
Fieser 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
Merck Index 15, 9831
Specific Gravity 0.728
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
Molecular Weight (g/mol) 101.19
Viscosity 0.36 mPa.s (20°C)
Formula Weight 101.19
Vapor Pressure 69mbar at 20°C
Percent Purity 99.7%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Triethylamine, 99.7%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'