missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylamine, 99.7%, extra pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
1 kg
2.5 kg
10 kg
Unit Size:
10kg
1kg
2.5kg
50g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -115.0°C |
| Density | 0.7280g/mL |
| Boiling Point | 90.0°C |
| Flash Point | −11°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.6% min. (GC) |
| Packaging | Plastic Bottle |
| Molecular Formula | C6H15N |
| Linear Formula | (C2H5)3N |
| Refractive Index | 1.3990 to 1.4010 |
| Show More |
Safety and Handling
Product Identifier
- Triethylamine
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Triethylamine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction