missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99%, pure, Thermo Scientific Chemicals

30.95 € - 252.00 €

Chemical Identifiers

CAS 121-44-8
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10046651
SDS View Documents
Thermo Scientific Acros
157910050
5 mL Glass bottle
30.95 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10651193
SDS View Documents
Thermo Scientific Acros
157911000
100 mL Glass bottle
32.27 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10265820
SDS View accessory products View Documents
Thermo Scientific Acros
157910010
1 L EcoSafPak™ Glass Bottle
44.63 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10626362
SDS View Documents
Thermo Scientific Acros
157910025
2.5 L Glass bottle
90.65 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11905181
SDS View Documents
Thermo Scientific Acros
157910100
10 L Plastic drum
252.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-44-8
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine

Specifications

121-44-8
0.7280g/mL
−11°C
98.5% min. (GC)
(C2H5)3N
04, 99
15, 9831
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
0.36 mPa.s (20°C)
69mbar at 20°C
Pure
Triethylamine
-115.0°C
90.0°C
Authentic
C6H15N
1.3990 to 1.4010
01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
0.728
Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
CCN(CC)CC
101.19
CHEBI:35026
101.19
99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-469-4

Product Identifier
  • Triethylamine

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.