Learn More
Triethylamine, 99+%, Thermo Scientific Chemicals
Beschreibung
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizers for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in the automotive casting industry and the textile industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
Melting Point | 115°C |
Density | 0.726 |
Boiling Point | 89°C to 90°C |
Flash Point | -11°C (12°F) |
Assay Percent Range | >99% |
Linear Formula | (CH3CH2)3N |
Refractive Index | 1.4005 |
Quantity | 250 g |
UN Number | UN1296 |
Beilstein | 1843166 |
Mehr anzeigen |
RUO – Research Use Only