missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, Extra Pure, SLR, Fisher Chemical™

25.60 € - 444.00 €

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number 9051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10615892
View Documents
Fisher Chemical
T/3200/08
500 mL Amber glass bottle
25.60 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11452984
View Documents
Fisher Chemical
T/3200/PB08
500 mL Plastic Coated Glass bottle
26.60 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10000490
View Documents
Fisher Chemical
T/3200/17
2.5 L Amber glass bottle
71.30 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10615702
View Documents
Fisher Chemical
T/3200/PB17
2.5 L Plastic Coated Glass bottle
69.75 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
12668655
View Documents
Fisher Chemical
T/3200/25
25 L Metal can
444.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

121-44-8
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine
C6H15N
9051
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC

Specifications

121-44-8
Colorless
90°C
9051
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
CHEBI:35026
69mbar at 20°C
-115°C
12.4
C6H15N
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CCN(CC)CC
101.193
3.5
101.19g/mol
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Triethylamine

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : Triethylamine

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.