Learn More
Triethylene glycol dimethacrylate, contains MEHQ as inhibitor, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 1.0920 g/mL |
Boiling Point | 170.0°C to 172.0°C (5.0 mmHg) |
Flash Point | 167°C |
Quantity | 250 mL |
Specific Gravity | 1.092 |
Solubility Information | Solubility aq. soln.: 3.6 g/L (20°C). Other solubilities: miscible with most organic solvents |
Viscosity | 11 mPa.s (25°C) |
Formula Weight | 286.32 |
Chemical Name or Material | Triethylene glycol dimethacrylate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling

- Triethylene glycol dimethacrylate, contains MEHQ as inhibitor
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Triethylene glycol dimethacrylate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.