Learn More
Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 175111000
138.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Triethyloxonium tetrafluoroborate | |
368-39-8, 75-09-2 | |
(C2H5)3OBF4 | |
01,1210; 02,430; 03,303; 04,527; 05,691; 06,611; 07,386; 08,500; 09,482; 10,417; 15,337 | |
triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium | |
IYDQMLLDOVRSJJ-UHFFFAOYSA-N | |
tetrafluoroboranuide; triethyloxidanium | |
2723982 | |
Plastic bottle | |
100 mL |
1M Solution in Methylene Chloride | |
C6H15BF4O | |
MFCD00044423 | |
1.3280g/mL | |
Solubility in water: reacts. | |
F[B-](F)(F)F.CC[O+](CC)CC | |
189.99 | |
189.99 | |
1.328 |
Safety and Handling
- Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only