Learn More
Triethylsilane, 98+%, Thermo Scientific Chemicals
32.56 € - 315.57 €
Chemical Identifiers
CAS | 617-86-7 |
---|---|
Molecular Formula | C6H15Si |
Molecular Weight (g/mol) | 115.271 |
MDL Number | MFCD00009018 |
InChI Key | QXTIBZLKQPJVII-UHFFFAOYSA-N |
Synonym | triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes |
PubChem CID | 6327258 |
IUPAC Name | triethylsilicon |
SMILES | CC[Si](CC)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484083
|
Thermo Scientific Alfa Aesar
A10320.09 |
10 g |
32.56 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494083
|
Thermo Scientific Alfa Aesar
A10320.18 |
50 g |
81.10 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404093
|
Thermo Scientific Alfa Aesar
A10320.30 |
250 g |
315.57 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, water and bases.
Chemical Identifiers
617-86-7 | |
115.271 | |
QXTIBZLKQPJVII-UHFFFAOYSA-N | |
6327258 | |
CC[Si](CC)CC |
C6H15Si | |
MFCD00009018 | |
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes | |
triethylsilicon |
Specifications
107°C to 108°C | |
617-86-7 | |
(CH3CH2)3SiH | |
UN1993 | |
−6°C (21°F) | |
Miscible with water. | |
CC[Si](CC)CC | |
115.271 | |
116.28 | |
1.412 | |
0.728 |
-157°C | |
C6H15Si | |
MFCD00009018 | |
1098278 | |
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes | |
QXTIBZLKQPJVII-UHFFFAOYSA-N | |
triethylsilicon | |
6327258 | |
≥98% | |
Moisture sensitive | |
Triethylsilane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 210-535-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only