missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilane, 99%

Product Code. 10022750 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10022750

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-86-7 | C6H16Si | 116.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-86-7
Molecular Formula C6H16Si
Molecular Weight (g/mol) 116.28
InChI Key QXTIBZLKQPJVII-UHFFFAOYSA-N
Synonym triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
PubChem CID 6327258
IUPAC Name triethylsilicon
SMILES CC[Si](CC)CC

Specifications

Boiling Point 107.0°C to 108.0°C
Color Colorless
Physical Form Liquid
Quantity 25 g
Assay Percent Range 99%
Linear Formula (C2H5)3SiH
Beilstein 04,625
Fieser 01,1218; 02,433; 03,304; 04,530; 05,694; 12,482; 13,340; 14,322; 15,338; 16,356; 17,367
Flash Point −3°C
Merck Index 15,9841
Solubility Information Solubility in water: decomposes
Formula Weight 116.28
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4100 to 1.4120
Specific Gravity 0.72
Density 0.7200g/mL
Chemical Name or Material Triethylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethylsilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.