missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilanol, 97%, Thermo Scientific Chemicals

Product Code. 11322286
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-52-4
Molecular Formula C6H16OSi
Molecular Weight (g/mol) 132.278
MDL Number MFCD00042640
InChI Key WVMSIBFANXCZKT-UHFFFAOYSA-N
Synonym triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47
PubChem CID 69005
IUPAC Name triethyl(hydroxy)silane
SMILES CC[Si](CC)(CC)O

Specifications

Boiling Point 154°C
Quantity 1 g
UN Number UN1993
Beilstein 1732848
Flash Point 57°C (135°F)
Formula Weight 132.28
Percent Purity 97%
Odor Mild
Refractive Index 1.433
Density 0.86
Chemical Name or Material Triethylsilanol
Show More Show Less
compliance-icons
Product Identifier
  • Triethylsilanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Triethylsilanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.