missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10264710
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15.0°C
Boiling Point 72.0°C
Color Colorless
Assay Percent Range 0.99
Linear Formula CF3CO2H
Beilstein 02, II, 186
Fieser 01,1219; 02,433; 03,305; 04,530; 05,695; 06,613; 07,388; 08,503; 09,483; 10,418; 11,557; 12,529; 14,322; 15,338; 16,83; 17,340
Merck Index 15, 9847
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane
IUPAC Name trifluoroacetic acid
Formula Weight 114.02
Percent Purity 99%
Grade Extra Pure
Density 1.5350 g/mL
Quantity 100 g
Infrared Spectrum Conforms to Reference
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3
Vapor Pressure 107 mbar at 25°C
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'