Learn More
Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
31.46 € - 674.00 €
Chemical Identifiers
CAS | 76-05-1 |
---|---|
Molecular Formula | C2HF3O2 |
Molecular Weight (g/mol) | 114.02 |
MDL Number | MFCD00004169 |
InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
PubChem CID | 6422 |
ChEBI | CHEBI:45892 |
SMILES | OC(=O)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031550
|
Thermo Scientific Acros
139720250 |
25 g | Glass Bottle |
31.46 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10264710
|
Thermo Scientific Acros
139721000 |
100 g | Glass Bottle |
66.83 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10785981
|
Thermo Scientific Acros
139725000 |
500 g | Glass Bottle |
238.19 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10021550
|
Thermo Scientific Acros
139720010 |
1 kg | Glass Bottle |
376.88 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10348483
|
Thermo Scientific Acros
139720025 |
2.5 kg | Glass Bottle |
674.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
76-05-1 | |
114.02 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
6422 | |
OC(=O)C(F)(F)F |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
CHEBI:45892 |
Specifications
76-05-1 | |
72.0°C | |
0.99 | |
CF3CO2H | |
02, II, 186 | |
15, 9847 | |
Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane | |
OC(=O)C(F)(F)F | |
114.02 | |
CHEBI:45892 | |
99% | |
1.5350 g/mL | |
Liquid | |
107 mbar at 25°C |
-15.0°C | |
Colorless | |
C2HF3O2 | |
MFCD00004169 | |
01,1219; 02,433; 03,305; 04,530; 05,695; 06,613; 07,388; 08,503; 09,483; 10,418; 11,557; 12,529; 14,322; 15,338; 16,83; 17,340 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
2,2,2-trifluoroacetic acid | |
6422 | |
114.02 | |
Extra Pure | |
Conforms to Reference | |
1.3 | |
Trifluoroacetic acid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Trifluoroacetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H290-May be corrosive to metals.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only