missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 10723857
Click to view available options
Quantity:
0.5 mL
1 mL
2 mL
10 mL
50 mL
Packaging:
Amber Glass
Ampule
Ampules

Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL

Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL

  • Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
  • Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
  • Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance 0.003 AU max. at 254nm, 0.03 AU max. at 300nm, 0.06 AU max. at 230nm, 0.06 AU max. at 275nm, 0.18 AU max. at 220nm, 0.55 AU max. at 210nm
pH 0.45
Melting Point -15°C
Boiling Point 72°C
Color Colorless
Assay Percent Range ≥99.5 %
Linear Formula CF3COOH
Merck Index 15, 9847
Formula Weight 114.02
Percent Purity ≥99.5%
Grade OPTIMA™ LC-MS
Density 1.535g/cm³
Quantity 50 mL
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation 1ppm max.
Vapor Density 3.9
Vapor Pressure 107mbar 25
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • TRIFLUOROACETIC ACID
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Trifluoroacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'