missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Puriss. p.a., for HPLC, ≥99.0% (GC), Honeywell Fluka™
Puriss. p.a., for HPLC,≥99.0% (GC)
520.00 € - 1247.24 €
Chemical Identifiers
CAS | 76-05-1 |
---|---|
Molecular Formula | C2HF3O2 |
Molecular Weight (g/mol) | 114.02 |
MDL Number | MFCD00004169 |
InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
PubChem CID | 6422 |
ChEBI | CHEBI:45892 |
SMILES | OC(=O)C(F)(F)F |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
76-05-1 | |
114.02 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
6422 | |
OC(=O)C(F)(F)F |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
CHEBI:45892 |
Specifications
max. 0.025 at 320 nm, max. 0.03 at 300 nm, max. 0.04 at 290 nm, max. 0.05 at 280 nm, max. 0.1 at 270 nm, max. 0.9 at 260 nm | |
-15.4°C (literature) | |
100.0000% | |
CH(OC2H5)3 | |
UN2699 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
OC(=O)C(F)(F)F | |
114.02 | |
CHEBI:45892 | |
>99% | |
1.489 g/mL at 20 °C(lit.) | |
Ampoule |
76-05-1 | |
72°C | |
C2HF3O2 | |
MFCD00004169 | |
742035 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
2,2,2-trifluoroacetic acid | |
6422 | |
114.02 | |
HPLC | |
100°C | |
Trifluoroacetic acid, Puriss. p.a., for HPLC, ≥99.0% (GC) |
Safety and Handling
missing translation for 'shelfLife' : 360 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: I; Proper Shipping Name: Trifluoroacetic acid
missing translation for 'einecsNumber' : 200-929-3
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Trifluoroacetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title