Learn More
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Description
Trifluoroacetic anhydride is used for the introduction of trifluoroacetyl group in organic synthesis. It is involved in the preparation of N- and O-trifluoroacetyl derivatives of a wide range of biologically active compounds for gas chromatography analysis. It plays an important role as desiccant for trifluoroacetic acid. It is also used in the oxidation of aldehydes to acids, esters, amides as well as in the protection of alcohols and amines. In addition, it is used as analytical reagents, solvents and dehydration condensing agent. It serves as an intermediate of fluorine fine chemicals, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trifluoroacetic anhydride is used for the introduction of trifluoroacetyl group in organic synthesis. It is involved in the preparation of N- and O-trifluoroacetyl derivatives of a wide range of biologically active compounds for gas chromatography analysis. It plays an important role as desiccant for trifluoroacetic acid. It is also used in the oxidation of aldehydes to acids, esters, amides as well as in the protection of alcohols and amines. In addition, it is used as analytical reagents, solvents and dehydration condensing agent. It serves as an intermediate of fluorine fine chemicals, pharmaceuticals and agrochemicals.
Solubility
Miscible with benzene, dichloromethane, diethyl ether, dimethylformamide, terahydrofuran and acetonitrile.
Notes
Hygroscopic and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, and alcohols.

Specifications
Specifications
Melting Point | -65°C |
Density | 1.503 |
Boiling Point | 39°C to 40°C |
Linear Formula | (CF3CO)2O |
Refractive Index | 1.269 |
Quantity | 25 g |
UN Number | UN3265 |
Beilstein | 746197 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with benzene,dichloromethane,diethyl ether,dimethylformamide,terahydrofuran and acetonitrile. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y22K032 | Certificate of Analysis | 11/12/2024 | A13614.22,A13614.14,A13614.36,A13614.0E |
Z20J047 | Certificate of Analysis | 10/01/2024 | A13614.22,A13614.14,A13614.36,A13614.0E |
10234904 | Certificate of Analysis | 06/02/2023 | A13614.22,A13614.14,A13614.36,A13614.0E |
10231997 | Certificate of Analysis | 06/02/2023 | A13614.22,A13614.14,A13614.36,A13614.0E |
M29H038 | Certificate of Analysis | 22/11/2021 | A13614.22,A13614.14,A13614.36,A13614.0E |
Safety and Handling


- Trifluoroacetic anhydride
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH014-Reacts violently with water.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Trifluoroacetic acidanhydride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.