Learn More
Trifluoromethanesulfonyl chloride, 98%, Thermo Scientific Chemicals
70.80 € - 739.00 €
Chemical Identifiers
CAS | 421-83-0 |
---|---|
Molecular Formula | CClF3O2S |
Molecular Weight (g/mol) | 168.51 |
MDL Number | MFCD00007451 |
InChI Key | GRGCWBWNLSTIEN-UHFFFAOYSA-N |
Synonym | trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl |
PubChem CID | 79000 |
IUPAC Name | trifluoromethanesulfonyl chloride |
SMILES | FC(F)(F)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420407
|
Thermo Scientific Alfa Aesar
B21774.06 |
5 g |
70.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430407
|
Thermo Scientific Alfa Aesar
B21774.14 |
25 g |
237.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440407
|
Thermo Scientific Alfa Aesar
B21774.22 |
100 g |
739.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRu(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.
Solubility
Hydrolyzes with water.
Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
421-83-0 | |
168.51 | |
GRGCWBWNLSTIEN-UHFFFAOYSA-N | |
79000 | |
FC(F)(F)S(Cl)(=O)=O |
CClF3O2S | |
MFCD00007451 | |
trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl | |
trifluoromethanesulfonyl chloride |
Specifications
421-83-0 | |
32°C | |
CClF3O2S | |
MFCD00007451 | |
1812016 | |
trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl | |
GRGCWBWNLSTIEN-UHFFFAOYSA-N | |
trifluoromethanesulfonyl chloride | |
79000 | |
98% |
1.593 | |
Pungent | |
1.334 | |
UN3265 | |
Moisture sensitive | |
Hydrolyzes with water. | |
FC(F)(F)S(Cl)(=O)=O | |
168.51 | |
168.52 | |
Trifluoromethanesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 207-009-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only