missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)trimethylsilane, 99%

Product Code. 10519453 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
1mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 10519453

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81290-20-2 | C5H11F3Si | 156.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81290-20-2
Molecular Formula C5H11F3Si
Molecular Weight (g/mol) 156.22
MDL Number MFCD00145454
InChI Key MTYSDPUZDZURPP-UHFFFAOYSA-N
Synonym trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
PubChem CID 552549
IUPAC Name trimethyl(trifluoromethyl)silane
SMILES CC(C)(C)[SiH2]C(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9500g/mL
Boiling Point 54.0°C to 55.0°C
Flash Point -17°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiCF3
Refractive Index 1.3300 to 1.3320
Quantity 25 mL
Merck Index 15, 9851
Specific Gravity 0.95
Formula Weight 142.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (Trifluoromethyl)trimethylsilane, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (Trifluoromethyl)trimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.