missing translation for 'onlineSavingsMsg'
Learn More

Triformamide, 97%, Thermo Scientific™

Product Code. 10403253
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 25891-31-0
MDL Number MFCD08059356
InChI Key PFBAGGWJGZAGCG-UHFFFAOYSA-N
Synonym triformamide
PubChem CID 6419719
IUPAC Name N,N-diformylformamide
SMILES C(=O)N(C=O)C=O

Specifications

Melting Point 100.0°C to 101.0°C
Color White
Elemental Analysis 96.0% min. (ex%C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H3NO3
Linear Formula N(COH)3
Quantity 1 g
Molecular Weight (g/mol) 101.06
Formula Weight 101.06
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material Triformamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'