missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triformamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Specifications
Specifications
Melting Point | 100.0°C to 101.0°C |
Color | White |
Elemental Analysis | 96.0% min. (ex%C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Molecular Formula | C3H3NO3 |
Linear Formula | N(COH)3 |
Quantity | 5 g |
Molecular Weight (g/mol) | 101.06 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'