Learn More
Manutan Trigger operated water gun, 150/10 psi/bar, 1/2 inch hose tail
Brand:  Manutan A327230
Encompass_Preferred
Product Code. 12803979
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Fingertip control of water usage in a variety of wash down applications. Adjustable spray/jet pattern with automatic water shut-off on release of trigger.
  • Working pressure : Max of 150psi
  • Working pressure : Max of 10bar
  • Max. flow rate : approx. 15L/min
  • 1/2 hose tail fitting