missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Triiodothyronine (T3) Competitive ELISA Kit

Competitive ELISA

Brand:  Invitrogen™ EIAT3C

 View more versions of this product

Product Code. 15835654

  • 610.00 € / 96 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, assay buffer, wash buffer, chromogen, stop solution, plate covers and lot-specific technical data sheet.
This item is not returnable. View return policy

Description

Description

The Triiodothyronine (T3) ELISA quantitates T3 in serum,plasma,urine,dried fecal extracts,or cell culture medium.

Principle of the method

The Triiodothyronine (T3) Competitive ELISA research-use-only kit is designed to quantitatively measure T3 present in extracted serum and plasma,urine,extracted dried fecal samples,and tissue culture media samples. This kit measures total T3 in extracted serum and plasma and in extracted fecal samples. A T3 stock solution is provided to generate standard curves for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture sheep antibodies. A T3-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a sheep antibody to T3 to each well. After a two-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound T3-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Thyroid hormones regulate a number of developmental, metabolic, and neural activities throughout the body. The thyroid gland synthesizes 2 hormones: Thyroxine, which contains 4 atoms of iodine (T4), and triiodothyronine (T3), which has 3 atoms of iodine. T3 production in the thyroid gland constitutes approximately 20% of the total T3; the rest is generated by the conversion (deiodination) of T4 to T3 in peripheral tissues. Thyroid hormones circulate primarily bound to carrier proteins (e.g., thyroidbinding globulin [TBG], prealbumin and albumin); whereas only a small fraction circulates unbound. The free form of T3 is the biologically active fraction. While both T3 and T4 are bound to TBG, T3 is bound less firmly than T4. Total T3 consists of both the bound and unbound fractions. Triiodothyronine (T3) is identical across all species.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

78.125-5000 pg/mL
Triiodothyronine (T3) Competitive ELISA Kit
HRP
ELISA Kit
Fecal Extract, Plasma, Serum, Supernatant, Urine
Human
Colorimetric Microplate Reader
6.3%
HRP
RUO
Fecal Extract,100 μL; Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL; Urine,25 μL
Chemical
2 hr. 30 min.
37.4 pg/mL
Multiplex assay on the Luminex Platform
Fluorescence
Thyroid hormone
96 assays
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
13.4%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Metabolism
-20°C
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.