missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropanolamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
1 kg
10 kg
Packaging:
Glass bottle
Plastic Can
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 48.0°C to 52.0°C |
Color | White to Yellow |
Boiling Point | 190.0°C (23.0 mmHg) |
Flash Point | 160°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Plastic Can |
Linear Formula | N(CH2CH(OH)CH3)3 |
Quantity | 10 kg |
Beilstein | 04,762 |
Show More |
Safety and Handling
Product Identifier
- Triisopropanolamine
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- H412-Harmful to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'