missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine cyclic borate, 98+%, Thermo Scientific Chemicals

Product Code. 10723275
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-00-8
Molecular Formula C9H18BNO3
Molecular Weight (g/mol) 199.06
MDL Number MFCD03791298
InChI Key IWKGJTDSJPLUCE-UHFFFAOYSA-N
Synonym triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
PubChem CID 225550
IUPAC Name 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane
SMILES B12OC(CN(CC(O1)C)CC(O2)C)C

Specifications

Melting Point 150°C to 157°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 500 g
Formula Weight 199.06
Percent Purity 98+%
Physical Form Crystalline Chunks and Needles
Chemical Name or Material Triisopropanolamine cyclic borate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'